Tin(II) oxalate, Thermo Scientific Chemicals
Tin(II) oxalate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tin(II) oxalate, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number 014113.36
also known as 014113-36
Price (USD)/ Each
120.00
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
120.00
Add to cart
Tin(II) oxalate, Thermo Scientific Chemicals
Catalog number014113.36
Price (USD)/ Each
120.00
-
Add to cart
Chemical Identifiers
CAS814-94-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay from Supplier's CofATin (Sn): ≥55.7%
Tin(II) oxalate is used for dyeing and printing textiles. It acts as a catalyst for the esterification reactions between long chain fatty acids and alcohols and in polymerization reactions. It is also used in stannous oral care compositions. Further, it is used in proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only