Thermo Scientific™

Tri-n-butylphosphine, 94%, Thermo Scientific Chemicals

Catalog number: 014114.95
2 x 500 g, Each
Thermo Scientific™

Tri-n-butylphosphine, 94%, Thermo Scientific Chemicals

Catalog number: 014114.95
2 x 500 g, Each
Quantity
Catalog number: 014114.95
also known as 014114-95
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
998-40-3
IUPAC Name
tributylphosphane
Molecular Formula
C12H27P
InChI Key
TUQOTMZNTHZOKS-UHFFFAOYSA-N
SMILES
CCCCP(CCCC)CCCC
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥92.5%

Description

Tri-n-butylphosphine is used as a catalyst modifier in the cobalt-catalyzed hydroformylation of alkenes for industrial use. It is also used as a strong reducing agent and polymerization cross-linking catalyst. It is a used as an intermediate for phase transfer catalysts. Used with platinum (II) or (IV) in an intermolecular hydroamination of inactivated alkenes. It is used as 1,4-addition catalyst; used with disulfides for the thioetherification of alcohols; acylation catalyst; used to prepare active esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • For cleavage of disulfide bonds, as a polymerization crosslinking catalyst, as an organic intermediate, and in fuels
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text