Thermo Scientific Chemicals

Tetra-n-butyltin, 95%, Thermo Scientific Chemicals

Catalog number: 014115.36
500 g, Each
Thermo Scientific Chemicals

Tetra-n-butyltin, 95%, Thermo Scientific Chemicals

Catalog number: 014115.36
500 g, Each
Quantity
Catalog number: 014115.36
also known as 014115-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1461-25-2
IUPAC Name
tetrabutylstannane
Molecular Formula
C16H36Sn
InChI Key
AFCAKJKUYFLYFK-UHFFFAOYSA-N
SMILES
CCCC[Sn](CCCC)(CCCC)CCCC
Form
Liquid
Appearance (Color)
Pale yellow
Assay from Suppliers CofA
≥94%

Description

Tetra-n-butyltin is used in hydrogenolysis reactions and as a transition metal catalyst. It is also used stabilizers in the manufacture of polyvinyl chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text