Di-n-butyltin dichloride, 96%, Thermo Scientific Chemicals
Di-n-butyltin dichloride, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Di-n-butyltin dichloride, 96%, Thermo Scientific Chemicals

Organotin intermediate
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
1 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 014116.30
also known as 014116-30
Price (USD)/ Each
74.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
87.70 
Save 13.05 (15%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
74.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
87.70 
Save 13.05 (15%)
Add to cart
Di-n-butyltin dichloride, 96%, Thermo Scientific Chemicals
Catalog number014116.30
Price (USD)/ Each
74.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
87.70 
Save 13.05 (15%)
-
Add to cart
Chemical Identifiers
CAS683-18-1
Molecular FormulaC8H20Cl2Sn
InChI KeyGSCBVGBIRKBMAC-UHFFFAOYSA-N
SMILESCl.Cl.CCCC[SnH2]CCCC
Molecular Weight (g/mol)305.86
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)>95.0%
Organotin intermediate.Di-n-butyltin dichloride is used as an organotin intermediate. It serves as heat and light stabilizer for polyvinyl chloride plastics. It is also used in the preparation of methyl 2-(methoxycarbonylmethyl)-5-iodobenzene by reacting with 2,5-diiodobenzoic acid. It acts as an esterification catalyst and a veterinary vermicide and tapeworm remedy. Further, it is used as ion exchange agent, process regulators and processing aids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only