Copper rod, 6.35mm (0.25in) dia, 99.9% (metals basis), Thermo Scientific Chemicals
Copper rod, 6.35mm (0.25in) dia, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Copper rod, 6.35mm (0.25in) dia, 99.9% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 cm
5 x 100 cm
Catalog number 014126.BY
also known as 014126-BY
Price (USD)/ Each
91.20
-
Add to cart
Quantity:
100 cm
Request bulk or custom format
Price (USD)/ Each
91.20
Add to cart
Copper rod, 6.35mm (0.25in) dia, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number014126.BY
Price (USD)/ Each
91.20
-
Add to cart
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.9% (metals basis)
Appearance (Color)Brown
FormRod, 5mm (0.2in) diameter.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only