Zinc fluoride tetrahydrate, 98%, Thermo Scientific Chemicals
Zinc fluoride tetrahydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc fluoride tetrahydrate, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
500 g
Catalog number 014127.36
also known as 014127-36
Price (USD)/ Each
148.65
Online exclusive
165.00 
Save 16.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
148.65
Online exclusive
165.00 
Save 16.35 (10%)
Add to cart
Zinc fluoride tetrahydrate, 98%, Thermo Scientific Chemicals
Catalog number014127.36
Price (USD)/ Each
148.65
Online exclusive
165.00 
Save 16.35 (10%)
-
Add to cart
Chemical Identifiers
CAS13986-18-0
IUPAC Namezinc(2+) tetrahydrate difluoride
Molecular FormulaF2H8O4Zn
InChI KeyOEUUYPRXVRBIFO-UHFFFAOYSA-L
SMILESO.O.O.O.[F-].[F-].[Zn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)97.5% min
Zinc fluoride tetrahydrate is used as a mild fluorinating agent in the replacement of chlorine in halogenated hydrocarbons, in fluorophosphate and fluoride glass, and in the manufacture of fluoride glass optical fibers and optical transmitting glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only