Sodium fluorophosphate, Thermo Scientific Chemicals
Sodium fluorophosphate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium fluorophosphate, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
500 g
Catalog number 014129.36
also known as 014129-36
Price (USD)/ Each
558.65
Online exclusive
620.00 
Save 61.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
558.65
Online exclusive
620.00 
Save 61.35 (10%)
Add to cart
Sodium fluorophosphate, Thermo Scientific Chemicals
Catalog number014129.36
Price (USD)/ Each
558.65
Online exclusive
620.00 
Save 61.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10163-15-2
IUPAC Namedisodium phosphorofluoridate
Molecular FormulaFNa2O3P
InChI KeyBFDWBSRJQZPEEB-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]P([O-])(F)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)94% min
Sodium fluorophosphate is used as an anti-inflammatory, glucocorticoid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only