Tin slug, 11mm (0.43in) dia x 4mm (0.2in) thick, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Tin slug, 11mm (0.43in) dia x 4mm (0.2in) thick, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tin slug, 11mm (0.43in) dia x 4mm (0.2in) thick, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 Pc.
10 Pc.
Catalog number 014134.KD
also known as 014134-KD
Price (USD)/ Each
728.00
-
Add to cart
Quantity:
50 Pc.
Request bulk or custom format
Price (USD)/ Each
728.00
Add to cart
Tin slug, 11mm (0.43in) dia x 4mm (0.2in) thick, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Catalog number014134.KD
Price (USD)/ Each
728.00
-
Add to cart
Chemical Identifiers
CAS7440-31-5
IUPAC Nametin
Molecular FormulaSn
InChI KeyATJFFYVFTNAWJD-UHFFFAOYSA-N
SMILES[Sn]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silvery
FormSlug, 11mm (0.43in) diameter.
Assay from Supplier's CofA≥99.999% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only