Lead(II) titanium oxide, 99.5%, Thermo Scientific Chemicals
Lead(II) titanium oxide, 99.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lead(II) titanium oxide, 99.5%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 014139.36
also known as 014139-36
Price (USD)/ Each
155.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
183.00 
Save 27.35 (15%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
155.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
183.00 
Save 27.35 (15%)
Add to cart
Lead(II) titanium oxide, 99.5%, Thermo Scientific Chemicals
Catalog number014139.36
Price (USD)/ Each
155.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
183.00 
Save 27.35 (15%)
-
Add to cart
Chemical Identifiers
CAS12060-00-3
IUPAC Nameλ²-lead(2+) oxotitaniumbis(olate)
Molecular FormulaO3PbTi
InChI KeyNKZSPGSOXYXWQA-UHFFFAOYSA-N
SMILES[Pb++].[O-][Ti]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay from Supplier's CofATiO2: ≥25.0%
Appearance (Color)White to cream
Assay from Supplier's CofAPbO: ≥74.5%
Lead(II) titanium oxide is widely used in ceramic capacitor, ceramic filters, PTC thermal resistor, pressure sensitive resistor and the performance of transducer field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only