Thermo Scientific Chemicals

Lead(II) titanium oxide, 99.5%, Thermo Scientific Chemicals

Catalog number: 014139.A3
2 kg, Each
Thermo Scientific Chemicals

Lead(II) titanium oxide, 99.5%, Thermo Scientific Chemicals

Catalog number: 014139.A3
2 kg, Each
Quantity
Catalog number: 014139.A3
also known as 014139-A3
Price (USD)
Have Questions?

Chemical Identifiers

CAS
12060-00-3
IUPAC Name
λ²-lead(2+) oxotitaniumbis(olate)
Molecular Formula
O3PbTi
InChI Key
NKZSPGSOXYXWQA-UHFFFAOYSA-N
SMILES
[Pb++].[O-][Ti]([O-])=O
Form
Powder
Assay from Supplier's CofA
TiO2: ≥25.0%
Appearance (Color)
White to cream
Assay from Supplier's CofA
PbO: ≥74.5%

Description

Lead(II) titanium oxide is widely used in ceramic capacitor, ceramic filters, PTC thermal resistor, pressure sensitive resistor and the performance of transducer field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text