Thermo Scientific Chemicals

Zinc sulfate monohydrate, Zn 35.5%, Thermo Scientific Chemicals

Catalog number: 014160.A1
1 kg, Each
Thermo Scientific Chemicals

Zinc sulfate monohydrate, Zn 35.5%, Thermo Scientific Chemicals

Catalog number: 014160.A1
1 kg, Each
Quantity
Catalog number: 014160.A1
also known as 014160-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7446-19-7
IUPAC Name
zinc(2+) hydrate sulfate
Molecular Formula
H2O5SZn
InChI Key
RNZCSKGULNFAMC-UHFFFAOYSA-L
SMILES
O.[Zn++].[O-]S([O-])(=O)=O
Assay (unspecified)
Zn 35.5% min

Description

Zinc sulfate monohydrate is used in calico-printing, preserving wood and skins, electrolytes for zinc plating, bleaching paper and clarifying glue. It is also used as a chemical reagent, coagulant in the preparation of rayon, mordant in dyeing and as a zinc source in animal feeds. In medicine, it is used as an astringent and emetic. Further, it is a precursor to the pigment lithopone. In addition to this, it is used to supply zinc in fertilizers, agricultural sprays, in electrolytes for zinc plating and as a mordant in dyeing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text