Gold slug, 6.35mm (0.25in) dia x 6.35mm (0.25in) length, Premion™, 99.999% (metals basis), Thermo Scientific Chemicals
Gold slug, 6.35mm (0.25in) dia x 6.35mm (0.25in) length, Premion™, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Gold slug, 6.35mm (0.25in) dia x 6.35mm (0.25in) length, Premion™, 99.999% (metals basis), Thermo Scientific Chemicals

Form: approx. 3.35-3.60g/slug, approx. weight
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 Pc.
1 Pc.
Catalog number 014174.KJ
also known as 014174-KJ
Price (USD)/ Each
4,002.00
-
Add to cart
Quantity:
5 Pc.
Request bulk or custom format
Price (USD)/ Each
4,002.00
Add to cart
Gold slug, 6.35mm (0.25in) dia x 6.35mm (0.25in) length, Premion™, 99.999% (metals basis), Thermo Scientific Chemicals
Catalog number014174.KJ
Price (USD)/ Each
4,002.00
-
Add to cart
Specifications
Chemical Name or MaterialGold
ColorGolden
CAS7440-57-5
AppearanceGold colored slug
Recommended StorageAmbient temperatures
View more
Gold slugs are used in thermal evaporation and electrodes (quartz oscillator, light-emitting diodes, and laser diodes).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only