Copper(II) acetate monohydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals
Copper(II) acetate monohydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Copper(II) acetate monohydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2 kg
Catalog number 014177.36
also known as 014177-36
Price (USD)/ Each
104.65
Online exclusive
116.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
104.65
Online exclusive
116.00 
Save 11.35 (10%)
Add to cart
Copper(II) acetate monohydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals
Catalog number014177.36
Price (USD)/ Each
104.65
Online exclusive
116.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS6046-93-1
IUPAC Namecopper(2+) diacetate hydrate
Molecular FormulaC4H8CuO5
InChI KeyNWFNSTOSIVLCJA-UHFFFAOYSA-L
SMILESO.[Cu++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentCa 0.005%
Impurity contentFe 0.002%
Impurity contentNa 0.05%
Insoluble material0.01%
Assay (unspecified)98.0-102.0% C₄H₆CuO₄.H₂O
View more
Copper(II) acetate monohydrate is used in DNA extraction in biochemical applications. It is also used as a catalyst or oxidizing agent in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only