Aluminum foil, 0.25mm (0.01in) thick, annealed, 99% (metals basis), Si and Fe (combined) typically 1%, Thermo Scientific Chemicals
Aluminum foil, 0.25mm (0.01in) thick, annealed, 99% (metals basis), Si and Fe (combined) typically 1%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum foil, 0.25mm (0.01in) thick, annealed, 99% (metals basis), Si and Fe (combined) typically 1%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
30 x 100 cm
30 x 50 cm
30 x 500 cm
Catalog number 014196.CJ
also known as 014196-CJ
Price (USD)/ Each
95.00
-
Add to cart
Quantity:
30 x 100 cm
Request bulk or custom format
Price (USD)/ Each
95.00
Add to cart
Aluminum foil, 0.25mm (0.01in) thick, annealed, 99% (metals basis), Si and Fe (combined) typically 1%, Thermo Scientific Chemicals
Catalog number014196.CJ
Price (USD)/ Each
95.00
-
Add to cart
Chemical Identifiers
CAS7429-90-5
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver
FormFoil
Total Metal Impurities1% max.
Comment0.22 - 0.28mm thick
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only