Thermo Scientific Chemicals

Telluric acid dihydrate, 99+%, Thermo Scientific Chemicals

Catalog number: 014197.09
10 g, Each
Thermo Scientific Chemicals

Telluric acid dihydrate, 99+%, Thermo Scientific Chemicals

Catalog number: 014197.09
10 g, Each
Quantity
Catalog number: 014197.09
also known as 014197-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13451-14-4
IUPAC Name
telluric acid dihydrate
Molecular Formula
H6O6Te
InChI Key
VCYUGZHEVVXOPD-UHFFFAOYSA-N
SMILES
O.O.O[Te](O)(=O)=O
Assay (unspecified)
99.1% min

Description

Telluric acid dihydrate is used in the preparation of tellurium nanorods through hydrazine-driven reduction, which finds application in solar cells, thermoelectric and topological insulators. It is also involved in the preparation of tellurium metal by reacting with the solution of potassium thiocyanate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text