Telluric acid dihydrate, 99+%, Thermo Scientific Chemicals
Telluric acid dihydrate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Telluric acid dihydrate, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number 014197.18
also known as 014197-18
Price (USD)/ Each
189.65
Online exclusive
211.00 
Save 21.35 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
189.65
Online exclusive
211.00 
Save 21.35 (10%)
Add to cart
Telluric acid dihydrate, 99+%, Thermo Scientific Chemicals
Catalog number014197.18
Price (USD)/ Each
189.65
Online exclusive
211.00 
Save 21.35 (10%)
-
Add to cart
Chemical Identifiers
CAS13451-14-4
IUPAC Nametelluric acid dihydrate
Molecular FormulaH6O6Te
InChI KeyVCYUGZHEVVXOPD-UHFFFAOYSA-N
SMILESO.O.O[Te](O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99.1% min
Telluric acid dihydrate is used in the preparation of tellurium nanorods through hydrazine-driven reduction, which finds application in solar cells, thermoelectric and topological insulators. It is also involved in the preparation of tellurium metal by reacting with the solution of potassium thiocyanate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only