Thermo Scientific Chemicals

Lead(II,IV) oxide, 97% (metals basis), Thermo Scientific Chemicals

Catalog number: 014232.A7
5 kg, Each
Thermo Scientific Chemicals

Lead(II,IV) oxide, 97% (metals basis), Thermo Scientific Chemicals

Catalog number: 014232.A7
5 kg, Each
Quantity
Catalog number: 014232.A7
also known as 014232-A7
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1314-41-6
IUPAC Name
bicyclo[3.1.1]triplumboxan-1-yl
Molecular Formula
O4Pb3
InChI Key
XMFOQHDPRMAJNU-UHFFFAOYSA-N
SMILES
O1[Pb]2O[Pb]1O[Pb]O2
Appearance (Color)
Red
Form
Powder
Assay from Supplier's CofA
Pb₃O₄: ≥96.0%

Description

Lead(II,IV) oxide is used to prepare colorless glass, faience glaze, porcelain painting flux, iron and steel coatings, rubber pigment and in glass cement. It is also used in gas and steam pipes, storage batteries, writing on glass and to make lead peroxide and matches. It is associated with linseed oil and used as a thick, long-lasting anti-corrosive paint. It gives better water resistant properties by replacing magnesium oxide. Further, it is used in the manufacture of lead glass and rustproof primer paints. In addition to this, it acts as a pigment for primer paints for iron objects.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text