Thermo Scientific Chemicals

Sodium selenate decahydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 014239.22
100 g, Each
Thermo Scientific Chemicals

Sodium selenate decahydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 014239.22
100 g, Each
Quantity
Catalog number: 014239.22
also known as 014239-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10102-23-5
IUPAC Name
disodium decahydrate selenate
Molecular Formula
H20Na2O14Se
InChI Key
TYVNFQRMDPFYPC-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-][Se]([O-])(=O)=O
Total Metal Impurities
0.1% max.

Description

Sodium selenate decahydrate is used as a feed additive, as a mineral supplement, as analytical reagents and in horticultural applications. Further, it is used in manufacturing of glass to counter the color caused by iron impurities. It is also employed in the prevention of abnormal tissue growth. In addition to this, it is used as a micronutrient which prevents a deficiency of copper, manganese and selenium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text