Thermo Scientific Chemicals

Lead(II) acetate trihydrate, ACS, 99.0-103.0%, Thermo Scientific Chemicals

Catalog number: 014242.A3
2 kg, Each
Thermo Scientific Chemicals

Lead(II) acetate trihydrate, ACS, 99.0-103.0%, Thermo Scientific Chemicals

Catalog number: 014242.A3
2 kg, Each
Quantity
Catalog number: 014242.A3
also known as 014242-A3
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6080-56-4
IUPAC Name
λ²-lead(2+) diacetate trihydrate
Molecular Formula
C4H12O7Pb
InChI Key
MCEUZMYFCCOOQO-UHFFFAOYSA-L
SMILES
O.O.O.[Pb++].CC([O-])=O.CC([O-])=O
Impurity content
Ca 0.005%
Impurity content
Cl 5ppm
Impurity content
Cu 0.002%
Impurity content
Fe 0.001%
Impurity content
K 0.005%

Description

Lead(II) acetate trihydrate is used as a mordant in textile printing and dyeing, drier in paints, varnishes and as a water repellant. It is also used in cosmetics and to prepare other lead compounds. It serves as an ingredient in progressive types of hair coloring dyes. Further, it is used in the detection of poisonous gas hydrogen sulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text