Lead(II) nitrate, ACS, 99.0% min, Thermo Scientific Chemicals
Lead(II) nitrate, ACS, 99.0% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lead(II) nitrate, ACS, 99.0% min, Thermo Scientific Chemicals

For Use As An Oxidizer in Dye Industry, in Manufacturing of Mother-of-Pearl, As Mordant in Printing Textile
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 014243.36
also known as 014243-36
Price (USD)/ Each
60.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
71.40 
Save 10.75 (15%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
60.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
71.40 
Save 10.75 (15%)
Add to cart
Lead(II) nitrate, ACS, 99.0% min, Thermo Scientific Chemicals
Catalog number014243.36
Price (USD)/ Each
60.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
71.40 
Save 10.75 (15%)
-
Add to cart
Chemical Identifiers
CAS10099-74-8
IUPAC Nameλ²-lead(2+) dinitrate
Molecular FormulaN2O6Pb
InChI KeyRLJMLMKIBZAXJO-UHFFFAOYSA-N
SMILES[Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99.0% Pb(NO₃)₂ min
Impurity contentCl 0.001%
Impurity contentCu 0.002%
Impurity contentFe 0.001%
Impurity contentK 0.005%
View more
As an oxidizer in dye industry, in manufacturing of mother-of-pearl, as mordant in printing textile, as sensitizer in photographyEmployed in industrial applications such as heat stabilization in nylon and polyesters, coatings of photothermographic paper, gold cyanidation and pyrotechnics. Finds

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only