Thermo Scientific Chemicals

Sodium nitrite, ACS, 97% min, Thermo Scientific Chemicals

Catalog number: 014244.22
100 g, Each
Thermo Scientific Chemicals

Sodium nitrite, ACS, 97% min, Thermo Scientific Chemicals

Catalog number: 014244.22
100 g, Each
Quantity
Catalog number: 014244.22
also known as 014244-22
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7632-00-0
IUPAC Name
sodium nitrite
Molecular Formula
NNaO2
InChI Key
LPXPTNMVRIOKMN-UHFFFAOYSA-M
SMILES
[Na+].[O-]N=O
Chloride content
≤0.005%
Impurity content
Calcium,Ca: ≤0.01%
Impurity content
Heavy metals (as Pb): ≤0.001%
Impurity content
Iron, Fe: ≤0.001%
Assay (unspecified)
≥97.0% Sodium nitrite

Description

Sodium nitrite is used in many industrial applications like meat curing, coloring and preserving. It is used as a reagent in analytical chemistry, as an antidote in cyanide poisoning, as an electrolyte in electrochemical grinding, as a cooling solution in closed loop systems and as an additive in industrial greases. It finds application as a corrosion inhibitor as well as in the rubber industry. In metallurgy, it is used for phosphatizing and detinning. It acts as a precursor to diazo dyes, nitroso compounds and various organic compounds like pharmaceuticals. As a food additive, it is used to prevent botulism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text