Thermo Scientific Chemicals

Ammonium hydrogen carbonate, 99.0%, Thermo Scientific Chemicals

Catalog number: 014249.A1
1 kg, Each
Thermo Scientific Chemicals

Ammonium hydrogen carbonate, 99.0%, Thermo Scientific Chemicals

Catalog number: 014249.A1
1 kg, Each
Quantity
Catalog number: 014249.A1
also known as 014249-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1066-33-7
IUPAC Name
dizinc(2+) diammonium tricarbonate
Molecular Formula
C3H8N2O9Zn2
InChI Key
QOPSSYZLVIENBR-UHFFFAOYSA-J
SMILES
[NH4+].[NH4+].[Zn++].[Zn++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
Assay (unspecified)
99.0% min

Description

In cooling baths, fire extinguishing compounds, baking powder formulations, and in the manufacture of ceramics and pigments. Also used for compost treatment to accelerate decomposition.Ammonium hydrogen carbonate is used as a reagent in chemical industries. It is an active component used in the manufacturing of fire-extinguishing compounds, pharmaceuticals, dyes, pigments and fertilizers. It finds application in the plastic, ceramics, leather and rubber industries. It is also used in cooling baths, scale and gypsum removing agents, leavening agent for baked goods and a defatting agent for textiles. Further, it acts as a buffer used in the purification of chemicals. In addition to this, it is involved in the gel digestion of proteins by trypsin and mass spectrometric analysis of proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • In cooling baths, fire extinguishing compounds, baking powder formulations, and in the manufacture of ceramics and pigments
  • Also used for compost treatment to accelerate decomposition
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text