Silver rod, 12.7mm (0.5in) dia, hard, 99.9% (metals basis), Thermo Scientific Chemicals
Silver rod, 12.7mm (0.5in) dia, hard, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silver rod, 12.7mm (0.5in) dia, hard, 99.9% (metals basis), Thermo Scientific Chemicals

Form: Approx. 13.28g/cm; Temper: hard
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 cm
2 cm
Catalog number 014256.BS
also known as 014256-BS
Price (USD)/ Each
702.65
Online exclusive
780.00 
Save 77.35 (10%)
-
Add to cart
Quantity:
10 cm
Request bulk or custom format
Price (USD)/ Each
702.65
Online exclusive
780.00 
Save 77.35 (10%)
Add to cart
Silver rod, 12.7mm (0.5in) dia, hard, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number014256.BS
Price (USD)/ Each
702.65
Online exclusive
780.00 
Save 77.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-22-4
IUPAC Namesilver
Molecular FormulaAg
InChI KeyBQCADISMDOOEFD-UHFFFAOYSA-N
SMILES[Ag]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver
FormRod, 12.7mm (0.5in) diameter, hardened.
Assay from Supplier's CofA≥99.9% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only