Thermo Scientific Chemicals

Ammonium bromide, ACS, 99.0% min, Thermo Scientific Chemicals

Catalog number: 014257.36
500 g, Each
Thermo Scientific Chemicals

Ammonium bromide, ACS, 99.0% min, Thermo Scientific Chemicals

Catalog number: 014257.36
500 g, Each
Quantity
Catalog number: 014257.36
also known as 014257-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12124-97-9
IUPAC Name
ammonium bromide
Molecular Formula
BrH4N
InChI Key
SWLVFNYSXGMGBS-UHFFFAOYSA-N
SMILES
[NH4+].[Br-]
Assay (unspecified)
99.0% NH₄Br min
Residue on ignition
0.01%
Impurity content
BrO₃ 0.002%
Impurity content
Fe 5ppm
Impurity content
SO₄ 0.005%

Description

Corrosion inhibitors, manufacturing of photographic films, plates, paper, lithography, fireproofing of woodsAmmonium bromide is often used as an intermediate source of bromide ion in laboratory processes and pharmaceutical preparations. It is most commonly used as an additive in the preparation of photographic films, fireproofing products and corrosion inhibitors. Ammonium bromide is widely utilized in the manufacture of photographic chemicals and emulsions. It is also used in pharmaceutical preparations. It finds application in fireproofing of wood and lithography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text