Thermo Scientific Chemicals

Copper(II) bromide, 98+%, Thermo Scientific Chemicals

Catalog number: 014258.22
100 g, Each
Thermo Scientific Chemicals

Copper(II) bromide, 98+%, Thermo Scientific Chemicals

Catalog number: 014258.22
100 g, Each
Quantity
Catalog number: 014258.22
also known as 014258-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7789-45-9
IUPAC Name
copper(2+) dibromide
Molecular Formula
Br2Cu
InChI Key
QTMDXZNDVAMKGV-UHFFFAOYSA-L
SMILES
[Cu++].[Br-].[Br-]
Form
Crystalline
Assay (unspecified)
≥98.0%
Appearance (Color)
Black

Description

Copper(II) bromide is used in photographic processing as an intensifier and as a brominating agent in organic synthesis. It is also used in the coupling of o-iodophenols and aryl acetylenes avoiding the use of palladium. It finds application in the preparation of NK1/NK2 receptor antagonists used in the regulation of tachykinin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text