Potassium methoxide, 90+%, Thermo Scientific Chemicals
Potassium methoxide, 90+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium methoxide, 90+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number 014261.30
also known as 014261-30
Price (USD)/ Each
119.65
Online exclusive
133.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
119.65
Online exclusive
133.00 
Save 13.35 (10%)
Add to cart
Potassium methoxide, 90+%, Thermo Scientific Chemicals
Catalog number014261.30
Price (USD)/ Each
119.65
Online exclusive
133.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
CAS865-33-8
IUPAC Namepotassium methanolate
Molecular FormulaCH3KO
InChI KeyBDAWXSQJJCIFIK-UHFFFAOYSA-N
SMILES[K+].C[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White to yellow
Assay (unspecified)≥90.0% (total alkalinity)
Potassium methoxide is used as a chemical intermediate. It is also used in biodiesel production and in fuel additives. It acts as a catalyst for the preparation of methyl formate and an alkaline catalyst for dimethyl formamide. Further, it is used as versatile reagents in chemical reactions such as condensation, esterification, alkoxylation and etherification, Claisen condensation, Wolf-Kishner reduction and Stobbe reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only