Thermo Scientific Chemicals

Magnesium ethoxide, Mg 21-22%, Thermo Scientific Chemicals

Catalog number: 014264.A1
1 kg, Each
Thermo Scientific Chemicals

Magnesium ethoxide, Mg 21-22%, Thermo Scientific Chemicals

Catalog number: 014264.A1
1 kg, Each
Quantity
Catalog number: 014264.A1
also known as 014264-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2414-98-4
IUPAC Name
magnesium(2+) diethanolate
Molecular Formula
C4H10MgO2
InChI Key
XDKQUSKHRIUJEO-UHFFFAOYSA-N
SMILES
[Mg++].CC[O-].CC[O-]
Assay (Aqueous acid-base Titration)
% Mg 20 - 22%
Infrared spectrum
Matches

Description

Magnesium ethoxide is used as olefin polymerization catalyst carrier for polypropylene, polyethylene and precision ceramic materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text