Thermo Scientific Chemicals

Sodium chlorite, tech. nominally 80%, Thermo Scientific Chemicals

Catalog number: 014265.A9
10 kg, Each
Thermo Scientific Chemicals

Sodium chlorite, tech. nominally 80%, Thermo Scientific Chemicals

Catalog number: 014265.A9
10 kg, Each
Quantity
Catalog number: 014265.A9
also known as 014265-A9
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7758-19-2
IUPAC Name
sodium chlorite
Molecular Formula
ClNaO2
InChI Key
UKLNMMHNWFDKNT-UHFFFAOYSA-M
SMILES
[Na+].[O-][Cl]=O
Assay from Supplier's CofA
Hydrogen Peroxide: Report
Assay from Supplier's CofA
Sodium Chlorate: Report
Assay from Supplier's CofA
Sodium Chloride: Report
Assay from Supplier's CofA
Sodium Chlorite: 77.5-82.5%
Assay from Supplier's CofA
Sodium Hydroxide: Report

Description

Sodium chlorite is used in the in situ generation of chlorine dioxide for stripping of textiles, bleaching, pulp and paper industries. It acts as a disinfectant in water treatment plant and as a preservative in eye drops. It is also used in contact lens cleaning solution and for sanitizing air ducts. It is associated with zinc chloride and used as a component in therapeutic rinses, toothpastes, mouth sprays and gels. It is utilized for the synthesis of 4-oxo-2-alkenoic acids from alkyl furans. Further, it is involved as a reagent in Pinnick oxidation reaction to prepare carboxylic acid from aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text