Thermo Scientific Chemicals

Neodymium(III) chloride, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 014282.06
5 g, Each
Thermo Scientific Chemicals

Neodymium(III) chloride, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 014282.06
5 g, Each
Quantity
Catalog number: 014282.06
also known as 014282-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10024-93-8
IUPAC Name
neodymium(3+) trichloride
Molecular Formula
Cl3Nd
InChI Key
ATINCSYRHURBSP-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[Nd+3]
Appearance (Color)
Purple
Residual water
< 100 ppm
Total Rare Earth Oxide Impurities
0.01% max.
Form
Powder

Description

Neodymium(III) chloride is used as a precursor for the production of neodymium metal. It is used as a catalyst and accelerates polymerization of various dienes such as polybutylene, polybutadiene, and polyisoprene. It has luminescence property and is widely used as fluorescent label in organic molecules, thereby helping in easy tracking of the compound using fluorescence microscope during various physical and chemical reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text