Neodymium(III) chloride, ultra dry, 99.99% (REO), Thermo Scientific Chemicals
Neodymium(III) chloride, ultra dry, 99.99% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Neodymium(III) chloride, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number 014282.14
also known as 014282-14
Price (USD)/ Each
773.65
Online exclusive
859.00 
Save 85.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
773.65
Online exclusive
859.00 
Save 85.35 (10%)
Add to cart
Neodymium(III) chloride, ultra dry, 99.99% (REO), Thermo Scientific Chemicals
Catalog number014282.14
Price (USD)/ Each
773.65
Online exclusive
859.00 
Save 85.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10024-93-8
IUPAC Nameneodymium(3+) trichloride
Molecular FormulaCl3Nd
InChI KeyATINCSYRHURBSP-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Nd+3]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Total Rare Earth Oxide Impurities0.01% max.
Appearance (Color)Purple
Residual water< 100 ppm
Neodymium(III) chloride is used as a precursor for the production of neodymium metal. It is used as a catalyst and accelerates polymerization of various dienes such as polybutylene, polybutadiene, and polyisoprene. It has luminescence property and is widely used as fluorescent label in organic molecules, thereby helping in easy tracking of the compound using fluorescence microscope during various physical and chemical reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only