Magnesium iodide, ultra dry, 99.996% (metals basis), Thermo Scientific Chemicals
Magnesium iodide, ultra dry, 99.996% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Magnesium iodide, ultra dry, 99.996% (metals basis), Thermo Scientific Chemicals

Light Sensitive and Hygroscopic
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
Catalog number 014283.09
also known as 014283-09
Price (USD)/ Each
552.65
Online exclusive
614.00 
Save 61.35 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
552.65
Online exclusive
614.00 
Save 61.35 (10%)
Add to cart
Magnesium iodide, ultra dry, 99.996% (metals basis), Thermo Scientific Chemicals
Catalog number014283.09
Price (USD)/ Each
552.65
Online exclusive
614.00 
Save 61.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10377-58-9
IUPAC Namemagnesium(2+) diiodide
Molecular FormulaI2Mg
InChI KeyBLQJIBCZHWBKSL-UHFFFAOYSA-L
SMILES[Mg++].[I-].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Particle Size-10 Mesh
Total Metal Impurities0.004% max.
Appearance (Color)White
FormBeads
Magnesium iodide is used as reagents and intermediates in pharmaceuticals. It is useful in Baylis-Hillman reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only