Cesium bromide, ultra dry, 99.9% (metals basis), Thermo Scientific Chemicals
Cesium bromide, ultra dry, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cesium bromide, ultra dry, 99.9% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 014289.09
also known as 014289-09
Price (USD)/ Each
149.65
Online exclusive
166.00 
Save 16.35 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
149.65
Online exclusive
166.00 
Save 16.35 (10%)
Add to cart
Cesium bromide, ultra dry, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number014289.09
Price (USD)/ Each
149.65
Online exclusive
166.00 
Save 16.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7787-69-1
IUPAC Namecaesium(1+) bromide
Molecular FormulaBrCs
InChI KeyLYQFWZFBNBDLEO-UHFFFAOYSA-M
SMILES[Br-].[Cs+]
View more
SpecificationsSpecification SheetSpecification Sheet
FormBeads
Total Metal Impurities0.1% max.
Residual water< 100 ppm
Particle Size-10 Mesh
Appearance (Color)White
Cesium bromide is used as reagent, intermediate in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only