Copper(II) sulfide, ultra dry, 99.98% (metals basis), Thermo Scientific Chemicals
Copper(II) sulfide, ultra dry, 99.98% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Copper(II) sulfide, ultra dry, 99.98% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 014300.09
also known as 014300-09
Price (USD)/ Each
255.65
Online exclusive
284.00 
Save 28.35 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
255.65
Online exclusive
284.00 
Save 28.35 (10%)
Add to cart
Copper(II) sulfide, ultra dry, 99.98% (metals basis), Thermo Scientific Chemicals
Catalog number014300.09
Price (USD)/ Each
255.65
Online exclusive
284.00 
Save 28.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1317-40-4
IUPAC Namecopper(2+) sulfanediide
Molecular FormulaCuS
InChI KeyOMZSGWSJDCOLKM-UHFFFAOYSA-N
SMILES[S--].[Cu++]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Cu 64.4 - 68.5%
Appearance (Color)Black
FormPowder
Total Metal Impurities0.02% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only