Calcium carbide, Thermo Scientific Chemicals
Calcium carbide, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Calcium carbide, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 x 1 kg
1 kg
Catalog number 014308.D9
also known as 014308-D9
Price (USD)/ Each
260.00
-
Add to cart
Quantity:
5 x 1 kg
Request bulk or custom format
Price (USD)/ Each
260.00
Add to cart
Calcium carbide, Thermo Scientific Chemicals
Catalog number014308.D9
Price (USD)/ Each
260.00
-
Add to cart
Chemical Identifiers
CAS75-20-7
IUPAC Namecalcium acetylene
Molecular FormulaC2Ca
InChI KeyUIXRSLJINYRGFQ-UHFFFAOYSA-N
SMILES[Ca++].[C-]#[C-]
View more
SpecificationsSpecification SheetSpecification Sheet
FormLump
Assay (unspecified)Ca 46% min
Transition TemperatureGray to black
Calcium carbide is used as a desulfurizer, dehydrant of steel, fuel in steel making, powerful deoxidizer and as a source of acetylene gas. It is used as a starting material for the preparation of calcium cyanamide, ethylene, chloroprene rubber, acetic acid, dicyandiamide and cyanide acetate. It is used in carbide lamps, toy cannons such as the big-bang cannon and bamboo cannon. It is associated with calcium phosphide and used in floating, self-igniting naval signal flares. Further, it is involved in the reduction of copper sulfide to metallic copper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only