Potassium sulfate, ACS, 99.0% min, Thermo Scientific Chemicals
Potassium sulfate, ACS, 99.0% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium sulfate, ACS, 99.0% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
Catalog number 014311.36
also known as 014311-36
Price (USD)/ Each
61.65
Online exclusive
67.90 
Save 6.25 (9%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
61.65
Online exclusive
67.90 
Save 6.25 (9%)
Add to cart
Potassium sulfate, ACS, 99.0% min, Thermo Scientific Chemicals
Catalog number014311.36
Price (USD)/ Each
61.65
Online exclusive
67.90 
Save 6.25 (9%)
-
Add to cart
Chemical Identifiers
CAS7778-80-5
IUPAC Namedipotassium sulfate
Molecular FormulaK2O4S
InChI KeyOTYBMLCTZGSZBG-UHFFFAOYSA-L
SMILES[K+].[K+].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentCl 0.001%
Impurity contentNa 0.02%
pHof a 5% solution 5-8.5 at 25?
Impurity contentCa 0.01%
Impurity contentFe 5ppm
View more
Potassium sulfate is used in the production of fertilizers (i.e. providing potassium and sulfur), potassium alum, potassium carbonate, and glass. In analytical chemistry, it is used in the determination of nitrogen by Kjeldahl method and protein in foodstuffs. Also, it is used in the electrostatic effects on pKa values of amino acid residues in the staphylococcal nuclease. It is used in the separation of protein through electrophoresis and also serves as a laboratory reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only