Thermo Scientific Chemicals

Potassium sulfate, ACS, 99.0% min, Thermo Scientific Chemicals

Catalog number: 014311.A3
2 kg, Each
Thermo Scientific Chemicals

Potassium sulfate, ACS, 99.0% min, Thermo Scientific Chemicals

Catalog number: 014311.A3
2 kg, Each
Quantity
Catalog number: 014311.A3
also known as 014311-A3
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7778-80-5
IUPAC Name
dipotassium sulfate
Molecular Formula
K2O4S
InChI Key
OTYBMLCTZGSZBG-UHFFFAOYSA-L
SMILES
[K+].[K+].[O-]S([O-])(=O)=O
Assay (unspecified)
99.0% K₂SO₄ min
Impurity content
Mg 0.005%
Insoluble material
0.01%
Impurity content
Ca 0.01%
Impurity content
Fe 5ppm

Description

Potassium sulfate is used in the production of fertilizers (i.e. providing potassium and sulfur), potassium alum, potassium carbonate, and glass. In analytical chemistry, it is used in the determination of nitrogen by Kjeldahl method and protein in foodstuffs. Also, it is used in the electrostatic effects on pKa values of amino acid residues in the staphylococcal nuclease. It is used in the separation of protein through electrophoresis and also serves as a laboratory reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text