Potassium thiocyanate, ACS, 98.5% min, Thermo Scientific Chemicals
Potassium thiocyanate, ACS, 98.5% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium thiocyanate, ACS, 98.5% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 014318.22
also known as 014318-22
Price (USD)/ Each
48.65
Online exclusive
53.50 
Save 4.85 (9%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
48.65
Online exclusive
53.50 
Save 4.85 (9%)
Add to cart
Potassium thiocyanate, ACS, 98.5% min, Thermo Scientific Chemicals
Catalog number014318.22
Price (USD)/ Each
48.65
Online exclusive
53.50 
Save 4.85 (9%)
-
Add to cart
Chemical Identifiers
CAS333-20-0
IUPAC Namepotassium cyanosulfanide
Molecular FormulaCKNS
InChI KeyZNNZYHKDIALBAK-UHFFFAOYSA-M
SMILES[K+].[S-]C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentCl 0.005%
Impurity contentNa 0.005%
Impurity contentSO₄ 0.005%
Assay (unspecified)98.5% KSCN min
Impurity contentFe 2ppm
View more
Potassium thiocyanate is used in the preparation of ethylenesulfide, lead thiocyante, which is used in the convertion of acyl chloride to thiocyanates. It is also used in solid state NMR of heavy-metal nuclei which results in the preparation of several mercury-thiocyanate and selenocyanate compounds as potential models. It finds application in cosmetic and personal care product formulations as a skin conditioning agent. Also used in moisturizers and oral care products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only