Nickel wire, 1.0mm (0.04in) dia, annealed, 99.5% (metals basis), Thermo Scientific Chemicals
Nickel wire, 1.0mm (0.04in) dia, annealed, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel wire, 1.0mm (0.04in) dia, annealed, 99.5% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 m
10 m
Catalog number 014337.H2
also known as 014337-H2
Price (USD)/ Each
243.65
Online exclusive
271.00 
Save 27.35 (10%)
-
Add to cart
Quantity:
50 m
Request bulk or custom format
Price (USD)/ Each
243.65
Online exclusive
271.00 
Save 27.35 (10%)
Add to cart
Nickel wire, 1.0mm (0.04in) dia, annealed, 99.5% (metals basis), Thermo Scientific Chemicals
Catalog number014337.H2
Price (USD)/ Each
243.65
Online exclusive
271.00 
Save 27.35 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialNickel wire
Name Note1.0mm (0.04 in.) dia., Annealed, Temper: soft
CAS7440-02-0
Health Hazard 1H317-H351-H372
Health Hazard 2GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only