This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 014369.AP
also known as 014369-AP
Price (USD)/ Each
198.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
233.00
Save 34.35 (15%)
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
198.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
233.00
Save 34.35 (15%)
Add to cart
Arsenic, plasma standard solution, Specpure™ As 10,000μg/mL, Thermo Scientific Chemicals
Chemical Name or MaterialArsenic plasma standard solution
ConcentrationAs in 5% HNO3
Name NoteAs 10000μg/mL
Health Hazard 1H290-H314-H335-H350
Quantity500 mL
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.