Thermo Scientific Chemicals
Gadolinium, plasma standard solution, Specpure™ Gd 10,000μg/mL, Thermo Scientific Chemicals
Catalog number: 014373.AD
50 mL, Each
Thermo Scientific Chemicals
Gadolinium, plasma standard solution, Specpure™ Gd 10,000μg/mL, Thermo Scientific Chemicals
Catalog number: 014373.AD
50 mL, Each
Quantity
Catalog number: 014373.AD
also known as 014373-AD
Price (USD)
Quantity
-
Have Questions?
Concentration
10,000?g/mL ?1%
Starting Material
Gd2O3