Chemical Name or MaterialRubidium plasma standard solution
ConcentrationRb2CO3 in 5% HNO3
Name NoteRb 10000μg/mL
Health Hazard 1H290-H314-H332-H335
Quantity50 mL
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.