Thermo Scientific Chemicals
Zirconium, plasma standard solution, Specpure™ Zr 10,000μg/mL, Thermo Scientific Chemicals
Catalog number: 014386.AP
500 mL, Each
Thermo Scientific Chemicals
Zirconium, plasma standard solution, Specpure™ Zr 10,000μg/mL, Thermo Scientific Chemicals
Catalog number: 014386.AP
500 mL, Each
Quantity
Catalog number: 014386.AP
also known as 014386-AP
Price (USD)
Have Questions?
Concentration
10,000?g/mL ?1%
Starting Material
ZrCl2O
Description
Zirconium plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.