Thermo Scientific Chemicals
Gold, plasma standard solution, Specpure™ Au 10,000μg/mL, Thermo Scientific Chemicals
Catalog number: 014392.AP
500 mL, Each
Thermo Scientific Chemicals
Gold, plasma standard solution, Specpure™ Au 10,000μg/mL, Thermo Scientific Chemicals
Catalog number: 014392.AP
500 mL, Each
Quantity
Catalog number: 014392.AP
also known as 014392-AP
Price (USD)
Quantity
-
Have Questions?
Starting Material
Au
Concentration
10,000?g/mL ?1%
Description
Gold, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.