Sodium, plasma standard solution, Specpure™ Na 10,000μg/mL, Thermo Scientific Chemicals
Sodium, plasma standard solution, Specpure™ Na 10,000μg/mL, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium, plasma standard solution, Specpure™ Na 10,000μg/mL, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
Catalog number 014400.AD
also known as 014400-AD
Price (USD)/ Each
66.65
Online exclusive
74.00 
Save 7.35 (10%)
-
Add to cart
Quantity:
50 mL
Request bulk or custom format
Price (USD)/ Each
66.65
Online exclusive
74.00 
Save 7.35 (10%)
Add to cart
Sodium, plasma standard solution, Specpure™ Na 10,000μg/mL, Thermo Scientific Chemicals
Catalog number014400.AD
Price (USD)/ Each
66.65
Online exclusive
74.00 
Save 7.35 (10%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialSodium, plasma standard solution
Health Hazard 1H290-H314-H335
Quantity50 mL
Health Hazard 2Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
Health Hazard 3P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
View more
Sodium, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only