Thermo Scientific Chemicals
Aluminum, plasma standard solution, Specpure™ Al 10,000μg/mL, Thermo Scientific Chemicals
Catalog number: 014405.AP
500 mL, Each
Thermo Scientific Chemicals
Aluminum, plasma standard solution, Specpure™ Al 10,000μg/mL, Thermo Scientific Chemicals
Catalog number: 014405.AP
500 mL, Each
Quantity
Catalog number: 014405.AP
also known as 014405-AP
Price (USD)
Quantity
-
Have Questions?
Starting Material
Al
Concentration
10,000?g/mL ?1%
Description
Aluminum, plasma standard solution is used as a single-element standard solution for plasma emission spectrometry. It is also used as a standard solution in analytical chemistry and atomic absorption spectroscopy.