Chemical Name or MaterialBarium plasma standard solution
ConcentrationBa(NO3)2 in 5% HNO3
Name NoteBa 10000μg/mL
Health Hazard 1H290-H314-H335
Quantity50 mL
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.