Neodymium, plasma standard solution, Specpure™ Nd 10,000μg/mL, Thermo Scientific Chemicals
Neodymium, plasma standard solution, Specpure™ Nd 10,000μg/mL, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Neodymium, plasma standard solution, Specpure™ Nd 10,000μg/mL, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
50 mL
500 mL
Catalog number 014431.AE
also known as 014431-AE
Price (USD)/ Each
134.65
Online exclusive
149.00 
Save 14.35 (10%)
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
134.65
Online exclusive
149.00 
Save 14.35 (10%)
Add to cart
Neodymium, plasma standard solution, Specpure™ Nd 10,000μg/mL, Thermo Scientific Chemicals
Catalog number014431.AE
Price (USD)/ Each
134.65
Online exclusive
149.00 
Save 14.35 (10%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialNeodymium plasma standard solution
ConcentrationNd2O3 in 5% HNO3
Name NoteNd 10000μg/mL
Health Hazard 1H290-H314-H335
Quantity100 mL
View more
Neodymium, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only