Manganese(II) chloride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals
Manganese(II) chloride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Manganese(II) chloride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 014464.06
also known as 014464-06
Price (USD)/ Each
66.65
Online exclusive
74.30 
Save 7.65 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
66.65
Online exclusive
74.30 
Save 7.65 (10%)
Add to cart
Manganese(II) chloride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number014464.06
Price (USD)/ Each
66.65
Online exclusive
74.30 
Save 7.65 (10%)
-
Add to cart
Chemical Identifiers
CAS7773-01-5
IUPAC Namemanganese(2+) dichloride
Molecular FormulaCl2Mn
InChI KeyGLFNIEUTAYBVOC-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Mn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pink
FormBeads
Residual water< 100 ppm
Total Metal Impurities0.01% max.
Particle Size-10 Mesh
Manganese(II) chloride is used in formation of an active Mn(0) species useful for radical cyclization reactions. It is used as catalyst, raw material for paint drier and antiknock agent for gasoline. It iis also used in the production of pigments, dyes, car batteries, chemical reagents and trace elements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only