Specifications
Chemical Name or MaterialStrontium isopropoxide
CAS88863-33-6
Health Hazard 1H228-H252-H314-H335
Health Hazard 2GHS H Statement
H314-H318-H228-H252
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
Self-heating in large quantities
may catch fire.
Health Hazard 3P210-P235+P410-P240-P241-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
View more