Ammonium hydrogen sulfate, 99.9% (metals basis), Thermo Scientific Chemicals
Ammonium hydrogen sulfate, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium hydrogen sulfate, 99.9% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number 014485.22
also known as 014485-22
Price (USD)/ Each
88.30
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
88.30
Add to cart
Ammonium hydrogen sulfate, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number014485.22
Price (USD)/ Each
88.30
-
Add to cart
Chemical Identifiers
CAS7803-63-6
IUPAC Nameammonium hydrogen sulfate
Molecular FormulaH5NO4S
InChI KeyBIGPRXCJEDHCLP-UHFFFAOYSA-N
SMILES[NH4+].OS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Ammonium hydrogen sulfate is used as pharmaceutical intermediates and chemical reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only